ReadyProbes™ Tissue Autofluorescence Quenching Kit
ReadyProbes™ Tissue Autofluorescence Quenching Kit
Invitrogen™

ReadyProbes™ Tissue Autofluorescence Quenching Kit

The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes unwanted autofluorescence in FFPE and frozen tissues due to aldehyde fixation, red bloodRead more
Have Questions?
Catalog NumberQuantity
R376301 Kit
Catalog number R37630
Price (EUR)
176,65
Online Exclusive
190,00
Save 13,35 (7%)
Each
-
Add to cart
Quantity:
1 Kit
Price (EUR)
176,65
Online Exclusive
190,00
Save 13,35 (7%)
Each
Add to cart
The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes unwanted autofluorescence in FFPE and frozen tissues due to aldehyde fixation, red blood cells, and structural elements such as collagen and elastin. If the target signal is low enough, then this background signal can interfere with the interpretation of immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence(IF), and in situ hybridization (ISH) assays. The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes most kinds of autofluorescence, with the exception of autofluorescence from lipofuschin granules.

The ReadyProbes Tissue Autofluorescence Quenching Kit contains three components that are combined just before use. Combine components A, B, and C in a 1:1:1 ratio. Mix components A and B together before adding component C. The resulting solution is then applied to the tissue for 2–5 minutes at room temperature. This is done after the blocking, primary antibody, and secondary antibody steps, but before application of any counterstains like nuclear stains.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeBlocking Reagent
Quantity1 Kit
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Product LineReadyProbes™
Unit SizeEach
Contents & Storage
A kit containing 5 mL each ready-to-use (1X) solutions of reagent A, B, and C in three clear HDPE bottles.
  • Store at 2–8°C.
  • When stored as directed, product can be used for 3 months after receipt.One kit is sufficient to tre

    Share catalog number, name or link