Search Thermo Fisher Scientific
Catalog Number | Type | Quantity | Certifications/Compliance |
---|---|---|---|
R40079 | Safranin | Each | CE Marked |
R40059 | Safranin | 5/Pk. | CE Marked |
R40058 | Safranin | Each | CE Marked |
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.