Remel™ QC-Slide™ Gram Stain Control
Remel™ QC-Slide™ Gram Stain Control
Thermo Scientific™
Remel™ QC-Slide™ Gram Stain Control
Remel QC-Slide Gram Stain Control is a heat-fixed quality control slide used for Gram stain procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R4014245 Slide/Pack
R4014010 Slides/Pk.
Catalog number R40142
Price (USD)
-
Request A Quote
Quantity:
45 Slide/Pack
Ask our AI about this Product
Remel QC-Slide Gram Stain Control is a heat-fixed quality control slide used for Gram stain procedures.
Specifications
DescriptionGram Stain Control
FormatGlass Slide
Product TypeGram Stain Control
Quantity45 Slide/Pack
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link