Remel™ m ENDO Agar LES
Remel™ m ENDO Agar LES
Remel™ m ENDO Agar LES
Remel™ m ENDO Agar LES
Thermo Scientific™
Remel™ m ENDO Agar LES
Recommended for cultivation and enumeration of coliform bacteria from water by the membrane filtration method.
Have Questions?
Catalog NumberQuantity
R453942500 g
Catalog number R453942
Price (KRW)
-
Quantity:
500 g
Ask our AI about this Product
Cultivate and enumerate coliform bacteria from water by membrane filtration method using Thermo Scientific™ Remel™ m ENDO Agar LES (Dehydrated). Endo Medium was first proposed by Endo in 1904 and allowed differentiation between lactose fermenters and non-fermenters. The m ENDO Agar LES medium is prepared according to the Lawrence Experimental Station (LES) formulation of McCarthy, Delaney, and Grasso1 and used for the enumeration of coliform organisms in water2.It is recommended by the American Public Health Association (APHA) for the enumeration of coliforms in water, wastewater, and foods2,3.
Coliforms are indicator organisms used for assessing the effectiveness of water treatment and disinfection, and for monitoring the sanitary integrity of water and wastewater. They are aerobic or facultatively anaerobic, gram-negative, non-spore forming bacteria that can ferment lactose and produce acid and gas.
Use m ENDO Agar LES for enumeration of coliforms by membrane filtration.
  • Selective Medium: Sodium lauryl sulfate and sodium deoxycholate inhibit non-coliforms
  • Recommended by the American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wastewater and Compendium of Methods for the Microbiological Examination of Foods2,3
  • Easy to interpret: Coliforms form red colonies with metallic green sheen within 24 hrs when incubated at 35°C
This medium contains lactose as the carbohydrate source. Casein peptone and casein-meat polypeptone provide essential nutrients for the growth of bacteria. Yeast extract acts as a growth enhancer and supplies B-complex vitamins. Monopotassium and dipotassium phosphate serve as buffers whereas sodium chloride provides electrolytes and maintains osmotic equilibrium. Sodium lauryl sulfate and sodium deoxycholate are selective agents that inhibit the growth of gram-positive bacteria. Basic fuchsin is the pH indicator. Agar is the solidifying agent.
Lactose-fermentation produces acetaldehyde that reacts with the sodium sulfite and basic fuchsin to form red colonies. A metallic green sheen is observed when the organism produces aldehydes with the rapid fermentation of lactose. Non-lactose fermenting colonies are clear, colorless or light pink.
On m-Endo LES Agar, coliforms appear as red colonies with a metallic green sheen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Not intended for IVD use.
  1. McCarthy J.A., J.E. Delaney, R.J. Grasso 1961 Measuring Coliforms in Water, Water and Sewage Works, 108. 238-243.
  2. Eaton A.D., L.S. Clesceri, E.W. Rice, A.E. Greenberg. 2005, Standard Methods for the Examination of Water and Wastewater. 21st APHA, Washington, DC:
  3. Downes F.P. and K. Ito. 2001. Compendium of Methods for the Microbiological Examination of Foods. 4th APHA, Washington, D.C.
Specifications
Descriptionm ENDO Agar LES
FormPowder
Product TypeAgar
Quantity500 g
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link