Triple Sugar Iron Agar (TSI) Medium (Dehydrated)
Triple Sugar Iron Agar (TSI) Medium (Dehydrated)
Triple Sugar Iron Agar (TSI) Medium (Dehydrated)
Triple Sugar Iron Agar (TSI) Medium (Dehydrated)
Triple Sugar Iron Agar (TSI) Medium (Dehydrated)
Thermo Scientific™

Triple Sugar Iron Agar (TSI) Medium (Dehydrated)

Obtain fast and accurate results with a medium designed for the differentiation of microorganisms based on carbohydrate fermentation and hydrogen sulfide production.
Have Questions?
Catalog NumberFormQuantityFill VolumeManufacturing Origin
R454982Powder1/Pk.500 gUnited States
Catalog number R454982
Price (USD)
-
Request A Quote
Form:
Powder
Quantity:
1/Pk.
Fill Volume:
500 g
Manufacturing Origin:
United States
Ask our AI about this Product
Differentiate gram-negative enteric bacilli such as Enterobacteriaceae on the basis of dextrose, lactose and sucrose fermentation and hydrogen sulfide production using Thermo Scientific™ Triple Sugar Iron Agar (TSI) (Dehydrated).
The sucrose content in this medium permits the recognition and exclusion of sucrose-fermenting species. These organisms may ferment lactose slowly, or not at all, during the incubation period, but they attack sucrose readily. Some Proteus and other species may give similar reactions to salmonellae and shigellae, and it is necessary to distinguish them by their ability to hydrolyse urea. Triple Sugar Iron Agar should be used in parallel with Urea Broth or Urea Agar.

TSI Agar is available as dehydrated culture medium or in ready-to-use prepared media tubes and in formats that meet international industrial guidelines for microbiological testing.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Certifications/ComplianceAOAC, BAM, COMPF, IDF, USDA, USP, SMWW
ColorStraw
DescriptionTriple Sugar Iron Agar (TSI) Medium
FormPowder
Manufacturing OriginUnited States
Product TypeAgar
Fill Volume500 g
Packaging TypePlastic bottle with red cap
Quantity1/Pk.
Shelf Life2 Years
Unit SizeEach
Contents & Storage
20°C to 25°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
136629Certificate of AnalysisSep 25, 2024R454982
468754Certificate of AnalysisApr 16, 2024R454982
891077Certificate of AnalysisApr 16, 2024R454982
741062Certificate of AnalysisApr 16, 2024R454982
738935Certificate of AnalysisApr 16, 2024R454982
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection