Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
R4601255 | 5 Loops/Pack |
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Share catalog number, name or link