Culti-Loops™ Geotrichum candidum ATCC™ 34614™
Culti-Loops™ Geotrichum candidum ATCC™ 34614™
Thermo Scientific™
Culti-Loops™ Geotrichum candidum ATCC™ 34614™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog number R4603001
Price (USD)/ Each
-
Request A Quote
Product Type:
Culti-Loops Quality Control Organism
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®34614TM*
DescriptionGeotrichum candidum ATCC™ 34614™
FormatCulti-Loops
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassFungi
Test TypeQualitative QC
Packaging Type5 Loops/Package
Strain DesignationsNo others found
Unit SizeEach