Culti-Loops™ Staphylococcus epidermidis ATCC™ 12228™
Culti-Loops™ Staphylococcus epidermidis ATCC™ 12228™
Thermo Scientific™
Culti-Loops™ Staphylococcus epidermidis ATCC™ 12228™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46065005 Loops/Pack
Catalog number R4606500
Price (USD)
-
Request A Quote
Quantity:
5 Loops/Pack
Ask our AI about this Product
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®12228TM*
DescriptionStaphylococcus epidermidis ATCC™ 12228™
FormatCulti-Loops
Quantity5 Loops/Pack
Strain DesignationsFDA strain PCI 1199
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassCocci
Test TypeQualitative QC
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link