Culti-Loops™ Streptococcus pneumoniae ATCC™ 49619
Culti-Loops™ Streptococcus pneumoniae ATCC™ 49619
Thermo Scientific™

Culti-Loops™ Streptococcus pneumoniae ATCC™ 49619

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
카탈로그 번호수량
R46090155 Loops/Pack
카탈로그 번호 R4609015
제품 가격(KRW)
-
수량:
5 Loops/Pack
Ask our AI about this Product
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.

Ordering Information:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
사양
ATCC®49619TM*
설명Streptococcus pneumoniae ATCC™ 49619™
형식Culti-Loops
수량5 Loops/Pack
균주 명칭WDCM00175, 262 [CIP 104340]
제품 유형Culti-Loop
타겟 유기체 분류Cocci
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

문서 및 다운로드

Certificates | 증명서

Lot 번호Certificate TypeDateCatalog Number(s)
262007Certificate of Analysis2025년 6월 14일R4609015
247551Certificate of Analysis2025년 4월 13일R4609015
228711Certificate of Analysis2025년 3월 19일R4609015
217524Certificate of Analysis2025년 2월 18일R4609015
205277Certificate of Analysis2025년 1월 11일R4609015
시험 성적서 관련 5 개의 결과가 검색되었습니다.

Safety Data Sheets | 물질 안전 보건 자료

Scientific Resources

Product Information

카탈로그 번호, 제품명 또는 링크 공유