Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog Number
Quantity
R4716500
100 Tests/Kit
Catalog number R4716500
Price (USD)
-
Request A Quote
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus epidermidis ATCC™ 12228™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
No serial dilution required
Delivers less-than100 colony forming units (CFUs) in a single 0.1mL inoculum
10 tests/reconstituted vial; 100 tests/kit
Ready for rehydration
For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.