Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing 2 QC Set
Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing 2 QC Set
Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing 2 QC Set
Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing 2 QC Set
Thermo Scientific™
Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing 2 QC Set
Supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Have Questions?
Catalog NumberQuantity
R4721001Set of 2, 100 Tests/Set
Catalog number R4721001
Price (USD)
-
Request A Quote
Quantity:
Set of 2, 100 Tests/Set
Ask our AI about this Product
Thermo Scientific™ Quanti-Cult Plus Growth Promotion Testing set is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
Includes:
10 preserved organism vials (two per organism); 12 rehydration vials
Contents:
  • Aspergillus brasiliensis ATCC™ 16404™†
  • Bacillus subtilis ATCC™ 6633™†
  • Candida albicans ATCC™ 10231™†
  • Pseudomonas aeruginosa ATCC™ 9027™†
  • Staphylococcus aureus subsp. aureus ATCC™ 6538™†
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®16404TM*
DescriptionGrowth Promotion Testing 2 QC Set/5
FormatQuanti-Cult Plus
Product TypeCulti-Loops Set
Target Organism ClassOrganism Sets
Test TypeQuantitative QC
Packaging Type100 Test/Set
QuantitySet of 2, 100 Tests/Set
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
569093Certificate of AnalysisApr 16, 2024R4721001
357462Certificate of AnalysisApr 16, 2024R4721001
281765Certificate of AnalysisApr 15, 2024R4721001
122059Certificate of AnalysisApr 15, 2024R4721001
654139Certificate of AnalysisApr 09, 2024R4721001
5 results displayed, search above for a specific certificate
Scientific Resources
Product Information
Share catalog number, name or link
1x1 image pixel for data collection