Remel™ Easyplate Stacker
Remel™ Easyplate Stacker
Thermo Scientific™
Remel™ Easyplate Stacker
Thermo Scientific™ Remel Easyplate Stacker is for storing and carrying petri dishes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
R55204Blue
R55225Assorted
R55208Clear
R55202Yellow
R55200Red
Catalog number R55204
Price (USD)
-
Request A Quote
Color:
Blue
Ask our AI about this Product
Thermo Scientific™ Remel Easyplate Stacker is for storing and carrying petri dishes.
  • Holds up to 56 plates
  • Easy to use and carry
  • Identification strip on side panel
  • Helps prevent spills
  • Color coded
Specifications
Product TypeEasyplate Stacker
ColorBlue
HoldsDishes
Positions56
ShapeSquare
StackableYes
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection