Phadebact™ CSF Test is used for direct detection of capsular antigens of S. pneumoniae, H. influenzae Type b, N. meningitidis group A, B, C, Y, W135 and S. agalactiae in cerebrospinal fluid, serum and urine.
Phadebact™ is a registered trademark of MKL Diagnostics AB.