Search Thermo Fisher Scientific
Catalog Number | Description | Quantity |
---|---|---|
R671877 | E. coli possessing special pili which are antigenetically different from those of other E. coli | 2 mL |
R671884 | Toxigenic E. coli Pili Antisera K99 | 2 mL |
Denka Seiken Agglutinating Sera, Toxigenic E. coli Pili Antisera are for the detection of E. coli possessing special pili which are antigenetically different from those of other E. coli.