Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
R681001 | 20/Pk. |
Mitsubishi™ AnaeroPack-Anaero is used for cultivation of anaerobic microorganisms without the addition of water or use of a catalyst.
Share catalog number, name or link