Invitrogen™
293FT Cell Line
Catalog number: R70007
Have Questions?
Invitrogen™
293FT Cell Line
Catalog number: R70007
Have Questions?
Catalog Number
R70007
Unit Size
1 x 107 cells
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R700071 x 107 cells
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The 293FT Cell line is ideal for generating high-titer lentivirus using the ViraPower™ Lentiviral Expression System. The 293FT Cell Line is a fast-growing, highly transfectable clonal isolate derived from human embryonal kidney cells transformed with the SV40 large T antigen. When a ViraPower™ expression vector and the ViraPower™ Lentiviral Packaging Mix are co-transfected into 293FT cells, high levels of the viral RNA and the gag⁄pol and rev proteins required for packaging are produced.
Advantages
• Fast growing
• Highly transfectable
• Produces high viral titers
• Allows very high levels of protein
Key Features
• Presence of the SV40 large T antigen allows very high levels of protein to be expressed from vectors containing the SV40 origin. Higher, in fact, than other 293 cells or the T antigen-transformed COS cells classically used.
Kit includes
• 293FT Cell Line provided as a frozen stock in 1 ml of 90% complete media and 10% DMSO
Specifications
Cell Line
293FT
Species
Human
Media Recommendation
SFM (Serum Free Medium)
No. of Cells
1 x 10^7 Cells
Product Line
ViraPower™
Quantity
1 x 10^7 Cells
Contents & Storage
• 293FT Cell Line (Store in liquid nitrogen)