ESwab™
ESwab™

ESwab™

Expand testing capabilities and experience savings by replacing multiple products with ESwab™, a liquid transport swab system compatible with automated specimen processing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure Color
R723481Green
R723482Blue
R723480White
Catalog number R723481
Price (USD)
-
Request A Quote
Closure Color:
Green
Ask our AI about this Product

Expand testing capabilities and make specimen collection and preservation simple with the ESwab™ System. ESwab is a liquid-based, multipurpose open platform, collection and preservation system that maintains viability of aerobic, anaerobic and fastidious bacteria for up to 48 hours at refrigerator and room temperature. The multipurpose ESwab offers significant cost savings—use it for automated specimen processing, gram stains, traditional bacteriology culture, rapid antigen tests* and molecular assays*.

*Self-validation required

Collect, snap and elute with ESwab; that is all it takes to simplify and streamline specimen collection.

In the testing laboratory, the ESwab cap is unscrewed and removed, and the swab applicator stick is securely attached to the cap. This allows the operator to conveniently remove the swab and perform various microbiology analyses, using the tube cap as a handle to hold and manipulate the swab.

Automated Specimen Processing

  • A collection and transport system cleared by the FDA to maintain viability of aerobes, anaerobes and fastidious bacteria for up to 48 hours at room temperature and refrigerator temperature*
  • Polypropylene screw-cap tube with internal conical shape filled with 1mL Liquid Aimes preservation medium elutes the entire sample upon contact
  • The only liquid transport swab system which allows for automated specimen processing with the COPAN WASP™DT line of instruments

Cost Savings

  • Expands testing capabilities with identical aliquots of liquid sample suspension, allowing for multiple tests from the same specimen
  • Allows for significant cost savings by replacing multiple products with just one, eliminating the need to stock multiple types of swabs
  • Creates more consistent gram stains as the entire specimen elutes from the FLOQSwab™, creating a homogenous suspension for a more precise and even smear

*Product tested and validated in full compliance with CLSI M40-A: Quality Control of Microbiological Transport System standard, including Neisseria gonorrhoeae survival at 24 hours.

COPAN WASP™DT instrumentation and accessories and ESwab collection and transport products are available through Thermo Fisher Scientific in the U.S.A. only. Please contact your local sales representative for more information.

ESwab and WASP™DT are trademarks or registered trademarks of COPAN S.P.A. All other trademarks are property of Thermo Fisher Scientific and its subsidiaries.

Research Use Only Authorization required
Specifications
Closure ColorGreen
DescriptionESwab Minitip Flocked Collection Kit
For Use With (Application)Eye, Ear, Nasal Passages, Urogential Tracts
Quantity50 Kits/Pk.
Shaft MaterialPlastic
Storage Requirements5°C to 25°C
Tip MaterialNylon Fiber
Tip TypeFlocked Minitip
Volume (Metric)1 mL
FormatTube
Product TypeMinitip Flocked Collection Kit
Swab ConfigurationSingle Applicator
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection