Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
R8311008 | 20 Panels/Kit |
The RapID CB PLUS System is a qualitative micromethod employing conventional and chromogenic substrates for the identification of medically important coryneform bacteria and other irregular Gram-positive bacilli isolated from human clinical specimens.
*Two-hour incubation for RapID SS/u.
A clear plastic tray contains reagent impregnated wells, allowing simultaneuos inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Innoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.
After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a databse or through the use of a computer-generated Code Compedium (ERIC).
Requires:
RapID Spot Indole, (R8309002), and RapID Inoculation Fluid, (R8325106)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
A qualitative micromethod using enzyme reactions to identify cinical isolates of corynebacterium species and other irregular gram positive bacilli. Used in a diagnostic workflow to aid clinicians in treatment options for patients suspected of having bacterial infections. The device is not automated, is for professional use only and is not a companion diagnostic.
Download Thermo Scientific™ RapID™ ERIC™ Software and stay up-to-date with our latest updates by visiting our Biochemical Identification Software Downloads portal ›
Share catalog number, name or link