Ni-NTA Agarose is a nickel-charged affinity resin that can be used to purify recombinant proteins containing a polyhistidine (6xHis) sequence.Read more
Have Questions?
Change view
Catalog Number
Quantity
R90115
25 mL
R90101
10 mL
R90110
100 mL
3 Options
Catalog number R90115
Price (USD)
459.65
Online Exclusive
489.00
Save 29.35 (6%)
Each
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)
459.65
Online Exclusive
489.00
Save 29.35 (6%)
Each
Add to cart
Ni-NTA Agarose is a nickel-charged affinity resin that can be used to purify recombinant proteins containing a polyhistidine (6xHis) sequence. Proteins bound to the resin may be eluted with either low pH buffer or by competition with imidazole or histidine. One-step purification can be performed under both native and denaturing conditions. Ni-NTA uses the chelating ligand nitrilotriacetic acid (NTA) coupled to a cross-linked 6% agarose resin that is suitable for use in batch and gravity flow applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity25 mL
Stationary PhaseNi-NTA
Column TypeAffinity Column
FormSuspension
TypeAgarose
Unit SizeEach
Contents & Storage
The Ni-NTA resin is pre-charged and able to bind up to 50 mg of recombinant protein per 1 ml of resin. It is provided as a 50% slurry in 30% ethanol. The resin will appear blue in color when charged with Ni2+ . Store at +4°C. Ni-NTA resin is guaranteed stable for 6 months when properly stored.
Frequently asked questions (FAQs)
Can ProBond or Ni-NTA beads be used for large-scale preparations?
ProBond and Ni-NTA beads can be used in FPLC columns. However, the beads can only withstand low pressure (~43.5 psi max).