| REFGAS030111 | Full specifications | Each | 3-channel gas chromatograph with configured valve oven | TRACE 1310 GC (2) TCD detectors and (1) FID detector, auxiliary valve oven with (2) 10-port valves, (2) 6-port valves, 5 columns, and complete documentation package. System is fully assembled and factory tested. | Request A Quote |
Type | Refinery Gas Analyzer | Detection Mode | Thermal Conductivity and Flame Ionization | Column Capacity | 5 | Description | 3-channel gas chromatograph with configured valve oven | Includes | TRACE 1310 GC (2) TCD detectors and (1) FID detector, auxiliary valve oven with (2) 10-port valves, (2) 6-port valves, 5 columns, and complete documentation package. System is fully assembled and factory tested. | Function | Simultaneous analysis of one gas sample for Permanent Gases, CO, CO2, C1-C8, and full range hydrogen and helium | Detector Type | TCD and FID | Unit Size | Each |
|
| LHA216300011 also known as TMP00148 | Full specifications | Each | Single channel GC analyzer for determing light hydrocarbon in natural gas, LPG, and liquid hydrocarbon samples | TRACE 1310 GC with (1) Split/Splitless inlet, (1) FID detector, auxiliary valve oven with (1) 4-port valve, (2) 6-port valves, 3 columns, and complete documentation package. System is fully assembled and factory tested. | Request A Quote |
Type | Light Hydrocarbon Analyzer | Detection Mode | Flame Ionization | Column Capacity | 3 | Description | Single channel GC analyzer for determing light hydrocarbon in natural gas, LPG, and liquid hydrocarbon samples | Includes | TRACE 1310 GC with (1) Split/Splitless inlet, (1) FID detector, auxiliary valve oven with (1) 4-port valve, (2) 6-port valves, 3 columns, and complete documentation package. System is fully assembled and factory tested. | Function | Analysis of single sample gas, liquid under pressure (LPG), or liquid at STP for hydrocarbons C1-C5 with backflush or C1-C9 with no backflush | Detector Type | FID | Unit Size | Each |
|
| REFGAS030121 | Full specifications | Each | 3-channel gas chromatograph with configured valve oven | TRACE 1310 GC (2) TCD detectors and (1) FID detector, auxiliary valve oven with (2) 10-port valves, (3) 6-port valves, 6 columns, and complete documentation package. System is fully assembled and factory tested. | Request A Quote |
Type | Refinery Gas Analyzer | Detection Mode | Thermal Conductivity and Flame Ionization | Column Capacity | 6 | Description | 3-channel gas chromatograph with configured valve oven | Includes | TRACE 1310 GC (2) TCD detectors and (1) FID detector, auxiliary valve oven with (2) 10-port valves, (3) 6-port valves, 6 columns, and complete documentation package. System is fully assembled and factory tested. | Function | Simultaneous analysis of one gas sample for Permanent Gases, CO, CO2, C1-C5 with early regroup composite peak for C6+, and full range hydrogen and helium | Detector Type | TCD and FID | Unit Size | Each |
|
| REFGAS030131 | Full specifications | Each | 3-channel gas chromatograph with configured valve oven | TRACE 1310 GC (2) TCD detectors and (1) FID detector, auxiliary valve oven with (2) 10-port Hastelloy C valves , (2) 6-port Hastelloy C valves, 6 columns, and complete documentation package. System is fully assembled and factory tested. | Request A Quote |
Type | Refinery Gas Analyzer | Detection Mode | Thermal Conductivity and Flame Ionization | Column Capacity | 6 | Description | 3-channel gas chromatograph with configured valve oven | Includes | TRACE 1310 GC (2) TCD detectors and (1) FID detector, auxiliary valve oven with (2) 10-port Hastelloy C valves , (2) 6-port Hastelloy C valves, 6 columns, and complete documentation package. System is fully assembled and factory tested. | Function | Simultaneous analysis of one gas sample which contains sulfur species for permanent gases, CO, CO2 and H2S, C1-C5 with early regroup composite peak for C6+, and full range hydrogen and helium | Detector Type | TCD and FID | Unit Size | Each |
|
| REFGAS040141 | Full specifications | Each | 4-channel gas chromatograph with configured valve oven | TRACE 1310 GC (2) TCD detectors and (2) FID detectors, auxiliary valve oven with (2) 10-port valves, (4) 6-port valves, 8 columns, and complete documentation package. System is fully assembled and factory tested. | Request A Quote |
Type | Refinery Gas Analyzer | Detection Mode | Thermal Conductivity and Flame Ionization | Column Capacity | 8 | Description | 4-channel gas chromatograph with configured valve oven | Includes | TRACE 1310 GC (2) TCD detectors and (2) FID detectors, auxiliary valve oven with (2) 10-port valves, (4) 6-port valves, 8 columns, and complete documentation package. System is fully assembled and factory tested. | Function | Simultaneous analysis of one gas sample which contains sulfur species for permanent gases, CO, CO2 and H2S, C1-C5 with early regroup composite peak for C6+, and full range hydrogen and helium | Detector Type | TCD and FID | Unit Size | Each |
|
| REFGAS040151 | Full specifications | Each | 4-channel gas chromatograph with configured valve oven | TRACE 1310 GC (2) TCD detectors and (2) FID detector, auxiliary valve oven with (2) 10-port valves, (3) 6-port valves, (1) methanizer, 6 columns, and complete documentation package. System is fully assembled and factory tested. | Request A Quote |
Type | Refinery Gas Analyzer | Detection Mode | Thermal Conductivity and Flame Ionization | Column Capacity | 6 | Description | 4-channel gas chromatograph with configured valve oven | Includes | TRACE 1310 GC (2) TCD detectors and (2) FID detector, auxiliary valve oven with (2) 10-port valves, (3) 6-port valves, (1) methanizer, 6 columns, and complete documentation package. System is fully assembled and factory tested. | Function | Simultaneous analysis of one gas sample for permanent gases, CO, CO2, C1-C5 with early regroup composite peak for C6+, low level of CO and CO2 and full range hydrogen and helium | Detector Type | TCD and FID | Unit Size | Each |
|
| REFGAS040161 | Full specifications | Each | 4-channel gas chromatograph with configured valve oven | TRACE 1310 GC (2) TCD detectors, (1) FID detector, (1) FPD detector, auxiliary valve oven with (2) 10-port Hastelloy C valves, (4) 6-port Hastelloy C valves, 8 columns, and complete documentation package. System is fully assembled and factory tested. | Request A Quote |
Type | Refinery Gas Analyzer | Detection Mode | Thermal Conductivity, Flame Ionization, Flame Photometry | Column Capacity | 8 | Description | 4-channel gas chromatograph with configured valve oven | Includes | TRACE 1310 GC (2) TCD detectors, (1) FID detector, (1) FPD detector, auxiliary valve oven with (2) 10-port Hastelloy C valves, (4) 6-port Hastelloy C valves, 8 columns, and complete documentation package. System is fully assembled and factory tested. | Function | Simultaneous analysis of one gas sample which contains sulfur species for permanent gases, CO, CO2 and H2S, C1-C5 with early regroup composite peak for C6+, sulfur speciation and full range hydrogen and helium | Detector Type | TCD, FID, FPD | Unit Size | Each |
|