Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
RFMS2305A | 651 L |
Protect flammable materials, solvents and intermediates using Thermo Scientific™ Revco FMS High-Performance Laboratory Refrigerators. These high-performance refrigerators are designed to meet stringent US Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids.† These refrigerators are designed to maintain a safe environment while keeping valuable samples within a tight temperature range. Each is listed by UL as a special purpose refrigerator intended for flammable material storage in accordance with NFPA 45, 70 and 99.† 45, 70 and 99.†
Ordering Information
Temperature chart recorders, and 4-20 milliamp are available separately. Not Available Outside North America
Note:
† Products complying with the applicable requirements of NFPA 70, National Electric Code, NPFA 45, Fire Protection for Laboratories Using Chemicals, and NFPA 99 Health Care Facilities are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.
Share catalog number, name or link