Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Highest antigen sequence indentity to the following orthologs: Mouse (32%), Rat (32%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62747. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TCL1B (T Cell Leukemia/Lymphoma 1B) is a Protein Coding gene. Diseases associated with TCL1B include T-Cell Leukemia and Lymphoma. Among its related pathways are PI3K-Akt signaling pathway. An important paralog of this gene is MTCP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Oncogene TCL-1B; Oncogene TCL1B; SYN-1; Syncytiotrophoblast-specific protein; T-cell leukemia/lymphoma protein 1B; T-cell lymphoma/leukemia 1B; TCL1/ MTCP1-like 1; TCL1/MTCP1-like protein 1
Gene Aliases: SYN-1; TCL1; TCL1B; TML1
UniProt ID: (Human) O95988
Entrez Gene ID: (Human) 9623
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support