Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (56%), Rat (56%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58208. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DYDC2 encodes a member of a family of proteins that contains a DPY30 domain. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA36D19.6; DPY30 domain-containing protein 2; MGC16186; SPS1; SRML1
Gene Aliases: DYDC2
UniProt ID: (Human) Q96IM9
Entrez Gene ID: (Human) 84332
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support