Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (36%), Rat (36%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65567. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be a transcriptional corepressor with KLF4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ZFP296; Zinc finger protein 296; Zinc finger protein 342
Gene Aliases: ZFP296; ZNF296; ZNF342
UniProt ID: (Human) Q8WUU4
Entrez Gene ID: (Human) 162979
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support