Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65863. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Apr; 4-glucantransferase; amylo-1, 6-glucosidase, 4-alpha-glucanotransferase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferaseprovided; Glycogen debrancher; Glycogen debranching enzyme; glycogen debranching protein; Oligo-1
Gene Aliases: AGL; GDE
UniProt ID: (Human) P35573
Entrez Gene ID: (Human) 178
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support