Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (33%), Rat (33%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82807. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen nuclear dot 52 kDa protein; Calcium-binding and coiled-coil domain-containing protein 2; Nuclear domain 10 protein 52; Nuclear domain 10 protein NDP52; Nuclear dot protein 52
Gene Aliases: CALCOCO2; NDP52
UniProt ID: (Human) Q13137
Entrez Gene ID: (Human) 10241
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support