Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55338. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
RP11-82K18.3 is an aminotransferase that transaminates kynurenine to form kynurenic acid. Kynurenic acid is a metabolite of tryptophan.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cysteine conjugate beta lyase 2; cysteine conjugate-beta lyase 2; Cysteine-S-conjugate beta-lyase 2; KATIII; Kynurenine aminotransferase 3; Kynurenine aminotransferase III; Kynurenine--glyoxylate transaminase; Kynurenine--oxoglutarate transaminase 3; Kynurenine--oxoglutarate transaminase III; kynurenine-oxoglutarate transaminase 3; RP11-82K18.3
Gene Aliases: CCBL2; KAT3; KATIII; KYAT3
UniProt ID: (Human) Q6YP21
Entrez Gene ID: (Human) 56267
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support