Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56412. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
LIN9 belongs to the lin-9 family and acts as a tumor suppressor. It inhibits DNA synthesis. Its ability to inhibit oncogenic transformation is mediated through its association with RB1. LIN9 plays a role in the expression of genes required for the G1/S transition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta subunit-associated regulator of apoptosis; HuLin-9; lin-9 homolog; pRB-associated protein; Protein lin-9 homolog; rb related pathway actor; TUDOR gene similar protein; Type I interferon receptor beta chain-associated protein
Gene Aliases: BARA; BARPsv; Lin-9; LIN9; TGS; TGS1; TGS2
UniProt ID: (Human) Q5TKA1
Entrez Gene ID: (Human) 286826
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support