Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58065. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
RFX6 is a member of the regulatory factor X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-binding protein RFX6; Regulatory factor X 6; Regulatory factor X domain-containing protein 1; regulatory factor X, 6
Gene Aliases: dJ955L16.1; MTCHRS; MTFS; RFX6; RFXDC1
UniProt ID: (Human) Q8HWS3
Entrez Gene ID: (Human) 222546
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support