Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59101. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein transport protein Sec24C; SEC24 family member C; SEC24 family, member C; SEC24 related gene family, member C; SEC24-related protein C
Gene Aliases: KIAA0079; SEC24C
UniProt ID: (Human) P53992
Entrez Gene ID: (Human) 9632
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support