Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56128. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this protein has not been specifically defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Speriolin-like protein; Spermatogenesis and centriole-associated protein 1-like protein
Gene Aliases: C21orf56; SPATC1L
UniProt ID: (Human) Q9H0A9
Entrez Gene ID: (Human) 84221
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support