Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (25%), Rat (25%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58252. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Citrate synthase-lysine N-methyltransferase CSKMT, mitochondrial; CS-KMT; Methyltransferase-like protein 12, mitochondrial; Putative methyltransferase LOC751071, mitochondrial
Gene Aliases: CSKMT; METTL12; U99HG
UniProt ID: (Human) A8MUP2
Entrez Gene ID: (Human) 751071
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support