Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61959. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin-Tcf signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dvl-binding protein; hNkd; hNkd1; naked cuticle-1; Naked-1; Protein naked cuticle homolog 1
Gene Aliases: Naked1; NKD; NKD1; PP7246
UniProt ID: (Human) Q969G9
Entrez Gene ID: (Human) 85407
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support