Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (56%), Rat (56%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60005. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ICA1L is a protein coding gene and gene ontology (GO) annotation includes acrosomal vesicle; protein domain specific binding; spermatid development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyotr; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 14; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 15; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 14 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 15 protein; Ica69-related protein; islet cell autoantigen 1,69kDa-like; Islet cell autoantigen 1-like protein
Gene Aliases: ALS2CR14; ALS2CR15; ICA1L
UniProt ID: (Human) Q96Q32
Entrez Gene ID: (Human) 130026
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support