Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | For Use With (Equipment) | Holds | No. of Racks |
---|---|---|---|
RSK23SD4 | Forma 900, Forma 7000, TSE and TSD Series (23 cu. ft. models) | Boxes (100/5 Racks) | 5 |
RSK13SD4 | Forma 900, Forma 7000, TSE and TSD Series (13 cu. ft. models) | Boxes (48/4 Racks) | 4 |
RSK13SD5 | HERAfreeze HFU B Series, Revco ExF and DxF Series (13 cu. ft. models) | Boxes (48/4 Racks) | 4 |
RSK17SD4 | Forma 900, Forma 7000, TSE and TSD Series (17 cu. ft. models) | Boxes (80/4 Racks) | 4 |
RSK17SD5 | HERAfreeze HFU B Series, Revco ExF and DxF Series (17 cu. ft. models) | Boxes (80/4 Racks) | 4 |
RSK23SD5 | HERAfreeze HFU B Series, Revco ExF and DxF Series | Boxes (80/5 Racks) | 5 |
RSK300SD4 | Thermo Scientific TDE Series (14.9 cu.ft.) | Boxes (75/3 Racks) | 3 |
RSK300SD5 | Thermo Scientific TDE Series (14.9 cu.ft.) | Boxes (60/3 Racks) | 3 |
RSK400SD4 | Thermo Scientific TSX and TDE Series (19 cu. ft.) | Boxes (100/4 Racks) | 4 |
RSK400SD5 | Thermo Scientific TSX and TDE Series (19 cu. ft.) | Boxes (80/4 Racks) | 4 |
RSK500SD4 | Thermo Scientific TSX and TDE Series (24.1 cu. ft.) | Boxes (125/5 Racks) | 5 |
RSK500SD5 | Thermo Scientific TSX and TDE Series (24.1 cu. ft.) | Boxes (100/5 Racks) | 5 |
RSK600SD4 | Thermo Scientific TSX and TDE Series (28.8 cu. ft.) | Boxes (150/6 Racks) | 6 |
RSK600SD5 | Thermo Scientific TSX and TDE Series (28.8 cu. ft.) | Boxes (120/6 Racks) | 6 |
RSK700SD4 | Thermo Scientific TSX Series (33.5 cu. ft.) | Boxes (175/7 Racks) | 7 |
RSK700SD5 | Thermo Scientific TSX Series (33.5 cu. ft.) | Boxes (140/7 Racks) | 7 |
RSK28CFSD4 | Forma 900, Forma 7000, TSE and TSD Series (28 cu. ft. models) | Boxes (150/6 Racks) | 6 |
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.