Thermo Scientific™
21PlusHD Measurement and Control System for Extrusion
Catalog number: S-APP50-04
Have Questions?
Thermo Scientific™
21PlusHD Measurement and Control System for Extrusion
Catalog number: S-APP50-04
Have Questions?
The ability to produce flat rolls with uniform thickness characteristics is paramount to the success of film and sheet manufacturers. Uniform thickness is important for sheet thermoforming, while maintaining minimum barrier film layer thickness is critical for food and medical packaging. The Thermo Scientific™ 21PlusHD for Extrusion measurement system allows manufacturers of cast, co-extruded, biax film and sheet products to minimize the thickness variation of the web, shift targets to save raw materials and decrease scrap and start-up times, while producing high performance products.
Catalog Number
S-APP50-04
Unit Size
Each
Description
Biax Film Extrusion Application Package (Dual Frame)
Price (USD)
Full specifications
ApplicationsExtrusion Products
DescriptionBiax Film Extrusion Application Package (Dual Frame)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Split Bubble Foam/Film Application Package
Price (USD)
Full specifications
ApplicationsExtrusion Products
DescriptionSplit Bubble Foam/Film Application Package
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sheet Extrusion Application Package
Price (USD)
Full specifications
ApplicationsExtrusion Products
DescriptionSheet Extrusion Application Package
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Cast Film Extrusion Application Package
Price (USD)
Full specifications
ApplicationsExtrusion Products
DescriptionCast Film Extrusion Application Package
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Co-extrusion Application Package
Price (USD)
Full specifications
ApplicationsExtrusion Products
DescriptionCo-extrusion Application Package
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Biax Film Extrusion Application Package (Single Frame)
Price (USD)
Full specifications
ApplicationsExtrusion Products
DescriptionBiax Film Extrusion Application Package (Single Frame)
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
S-APP50-04Full specifications
EachBiax Film Extrusion Application Package (Dual Frame)Request A Quote
ApplicationsExtrusion Products
DescriptionBiax Film Extrusion Application Package (Dual Frame)
Unit SizeEach
S-APP50-00Full specifications
EachSplit Bubble Foam/Film Application PackageRequest A Quote
ApplicationsExtrusion Products
DescriptionSplit Bubble Foam/Film Application Package
Unit SizeEach
S-APP50-01Full specifications
EachSheet Extrusion Application PackageRequest A Quote
ApplicationsExtrusion Products
DescriptionSheet Extrusion Application Package
Unit SizeEach
S-APP50-02Full specifications
EachCast Film Extrusion Application PackageRequest A Quote
ApplicationsExtrusion Products
DescriptionCast Film Extrusion Application Package
Unit SizeEach
S-APP50-03Full specifications
EachCo-extrusion Application PackageRequest A Quote
ApplicationsExtrusion Products
DescriptionCo-extrusion Application Package
Unit SizeEach
S-APP50-05Full specifications
EachBiax Film Extrusion Application Package (Single Frame)Request A Quote
ApplicationsExtrusion Products
DescriptionBiax Film Extrusion Application Package (Single Frame)
Unit SizeEach
Showing 6 of 6
The 21PlusHD for Extrusion incorporates a suite of advanced application packages for film and sheet manufacturing to increase the quality, consistency and performance characteristics of cast film, cast sheet, co-extrusion, and biax film products.
Advanced sensor technology is at the heart of the 21PlusHD platform through the Thermo Scientific™ PROSIS full spectrum NIR, Thermo Scientific™ ShadowMaster LED laser, and the latest Thermo Scientific™ Beta Plus nucleonic sensor devices. 
Each application package is specifically designed for the different processes required in the manufacture of world-class extruded products.  The suite of Thermo Scientific advanced machine-direction and cross-direction controls improve both quality and productivity, delivering consistent results and an impressive return on investment. 
Automatic Profile Control (APC) uses advanced mapping strategies for both the linear and non-linear (neck-in) regions of the extruded product. Machine Direction (MD) thickness control strategies include controlling line speed, screw or pump speed, or extruder ratio, to maintain the extruded product to thickness specifications. Options for increasing output (Throughput Control) and minimizing resin usage (Target Management Control) are also available. Target Management Control adjusts product to a system-calculated dynamic target, providing both low point and low average protection.
The APC package includes hardware and software to control auto dies to achieve and maintain cross-direction uniformity of extruded film. APC utilizes high-resolution product profile measurements from the systems scanning sensors to continuously adjust die bolts, resulting in reduced profile variations. When coupled with MD Control, APC ensures product uniformity and superior roll quality, resulting in quality improvement, scrap reduction, and material savings for tangible economic returns.
  • Application package includes core operating software for film and sheet extrusion lines
  • Standard user-friendly displays configured specifically for extrusion applications
  • Extensive support for both mono and co-extrusion lines
  • Accommodates the full line of Thermo Scientific sensors for a variety of measurements.
  • Compatible with most major makes of line and die manufacturers.
  • Wide variety of reporting options including roll, slit roll, shift, product production.
  • Extensive link options to automation control systems.
  • Comprehensive suite of APC and MD control options
  • PROSIS non-nuclear thickness and weight measurement available
HMI Specifications:
  • Wonderware™ InTouch visualization software version 14 R2
  • Dell OptiPlex XE3 Pc
  • Windows™ 10 
Available Displays
  • 2D profiles
  • 3D contour view
  • Blanket view ( heat map display)
  • MD and CD controls
  • Trends
  • Edge and full zoom
  • Roll reports
  • Diagnostics
  • SPC/QC
  • Single point / FFT
  • Defect report
To help maintain optimum performance and minimize downtime, Thermo Scientific Instrument Performance Management (IPM) Software is included with a Product Support Agreement (PSA). IPM Software is a cloud-based service platform that remotely monitors the health, status and performance of gauging instruments. With advanced diagnostics and capabilities to intelligently identify critical issues, it delivers a convenient way for customers to work. Notifications are automatically sent to our Technical Support Team to initiate troubleshooting and faster repair times. IPM Software also provides process analytics, data trending, historical data retrieval and data loss prevention to support quality management activities such as root cause analysis, traceability and auditing. In addition, customers have access to insights on operation costs and ROI on service contracts to help improve overall equipment effectiveness, reduce operating costs, and maximize return on investment.