Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
Applications | Nonwoven Materials |
Description | Nonwoven |
Unit Size | Each |
Applications | Nonwoven - Blend Measurement |
Description | Nonwoven – Blend Measurement |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (HKD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
S-APP50-20 | Each | Nonwoven | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S-APP50-21 | Each | Nonwoven – Blend Measurement | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The 21PlusHD Measurement and Control System for Nonwovens consists of a Mark III or L400 scanner, a Thermo Scientific™ PROSIS™ Infrared thickness sensor or Thermo Scientific™ Beta Plus™ sensor, binder and moisture measurements, and line speed controls.
Operator Interface:
Features WonderWare HMI user interface. Operator stations are PC-based and communicate with other system modules across an Ethernet LAN. Desktop interface and LCD450-console configurations are available.
Communication Capabilities:
The 21PlusHD system provides powerful communication capabilities with industry-accepted OPC standards, allowing the sharing of product and process data with popular software packages for data acquisition, database building and quality analysis.
Instrument Performance Management:
Maintain the health, status and performance of gauging instruments with Thermo Scientific Instrument Performance Management (IPM) Software, which is included as part of a Product Support Agreement (PSA). This cloud-based, 24/7 remote monitoring platform provides automatic notifications for troubleshooting and advanced diagnostics for fast restoration of performance and minimize downtime.