Human Keratinocyte Growth Supplement (HKGS)
Human Keratinocyte Growth Supplement (HKGS)
Gibco™

Human Keratinocyte Growth Supplement (HKGS)

Human Keratinocyte Growth Supplement (HKGS) is a sterile, concentrated (100X) solution intended for use with EpiLife medium or Medium 154Read more
Have Questions?
Catalog NumberQuantity
S00155 mL
Catalog number S0015
Price (USD)
69.65
Online Exclusive
74.75
Save 5.10 (7%)
Each
Add to cart
Quantity:
5 mL
Price (USD)
69.65
Online Exclusive
74.75
Save 5.10 (7%)
Each
Add to cart
Human Keratinocyte Growth Supplement (HKGS) is a sterile, concentrated (100X) solution intended for use with EpiLife medium or Medium 154 to culture human epidermal keratinocytes.

HKGS is an ionically-balanced supplement containing bovine pituitary extract (BPE), recombinant human insulin-like growth factor-I, hydrocortisone, bovine transferrin, and human epidermal growth factor. Each 5-mL bottle of HKGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human epidermal keratinocytes and is the correct amount of supplement for a 500-mL bottle of medium.

When a 500-mL bottle of medium is supplemented with HKGS, the final concentrations of the components in the supplemented medium are:
• Bovine pituitary extract (BPE): 0.2% v/v
• Recombinant human insulin-like growth factor-I: 0.01 μg/mL
• Hydrocortisone: 0.18 μg/mL
• Bovine transferrin: 5 μg/mL
• Human epidermal growth factor: 0.2 ng/mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeKeratinocyte Cells
Concentration100 X
For Use With (Application)Mammalian Cell Culture, Primary Cell Culture
Product LineCascade Biologics™
Quantity5 mL
SourceHuman
SpeciesHuman
FormLiquid
Product TypeGrowth Supplement
SterilitySterile
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C).

Frequently asked questions (FAQs)

Do I need to add additional supplements to AmnioMAX-II Complete Medium (Cat. No. 11269016)?

No. AmnioMax-II Complete Medium (Cat. No. 1129016) is fully supplemented with antibiotics, L-glutamine, and FBS.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link