Informational:Find out about our new and improved Large Vessel Endothelial Supplement (LVES).
Gibco™

Low Serum Growth Supplement (LSGS)

Catalog number: S00310
Have Questions?
Gibco™

Low Serum Growth Supplement (LSGS)

Catalog number: S00310
Have Questions?
Catalog Number
S00310
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S0031010 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Low Serum Growth Supplement (LSGS) is a sterile, concentrated (50X) solution intended for use as one component in a complete culture environment for the growth of human dermal fibroblasts and endothelial cells derived from large vessels. LSGS is intended for use with Medium 106 (fibroblasts) or Medium 200 (endothelial cells).

LSGS contains: fetal bovine serum, basic fibroblast growth factor, heparin, hydrocortisone, and epidermal growth factor.

When a 500-mL bottle of medium is supplemented with LSGS or the components of the LSGS Kit, the final concentrations of the components in the supplemented medium are:
• Fetal bovine serum: 2% v/v
• Hydrocortisone: 1 μg/mL
• Human epidermal growth factor: 10 ng/mL
• Basic fibroblast growth factor: 3 ng/mL
• Heparin: 10 μg/mL
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Epithelial Cells (Large Vessel), Fibroblast Cells (Dermal)
Concentration
50 X
Product Type
Growth Supplement
Sterility
Sterile
Product Line
Cascade Biologics™
Shipping Condition
Dry Ice
Species
Human

Contents & Storage

Store in freezer (-5 to -20°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text