Gibco™

Low Serum Growth Supplement Kit (LSGS Kit)

Catalog number: S003K
Have Questions?
Gibco™

Low Serum Growth Supplement Kit (LSGS Kit)

Catalog number: S003K
Have Questions?
Catalog Number
S003K
Unit Size
1 Kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S003K1 Kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The LSGS Kit provides, in separate vials, all the components of complete LSGS (S-003-10): fetal bovine serum, hydrocortisone, human epidermal growth factor, and basic fibroblast growth factor/heparin (stabilized with BSA). A vial of gentamycin/amphotericin solution (GA) is also included. Use of GA is optional. When a 500 ml bottle of medium is supplemented with LSGS or the components of the LSGS Kit, the final concentrations of the components in the supplemented medium are:
• fetal bovine serum, 2% v/v
• hydrocortisone (1 μg/ml)
• human epidermal growth factor, 10 ng/ml
• basic fibroblast growth factor, 3 ng/ml
• heparin, 10 μg/ml
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Epithelial Cells (Large Vessel), Fibroblast Cells (Dermal)
Product Type
Growth Supplement
Sterility
Sterile
Product Line
Cascade Biologics™
Quantity
1 Kit
Shipping Condition
Dry Ice
Species
Human
Unit Size
1 Kit

Contents & Storage

minus 20 degrees C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text