Gibco™

Microvascular Growth Supplement (MVGS)

Catalog number: S00525
Have Questions?
Gibco™

Microvascular Growth Supplement (MVGS)

Catalog number: S00525
Have Questions?
Catalog Number
S00525
Unit Size
25 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S0052525 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
A sterile, concentrated (20X) solution intended for use with Medium 131 to culture human microvascular endothelial cells. Each 25 ml bottle of MVGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human microvascular endothelial cells, and is the correct amount of supplement for a 500 ml bottle of Medium 131. When a 500 ml bottle of medium is supplemented with MVGS, the final concentrations of the components in the supplemented medium are:
• fetal bovine serum (4.9% v/v final concentration)
• hydrocortisone (1 μg/ml)
• human fibroblast growth factor (3 ng/ml)
• heparin (10 μg/ml)
• human epidermal growth factor (1 ng/ml)
• dibutyryl cyclic AMP (0.08 mM)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
20 X
Product Type
Growth Supplement
Sterility
Sterile
Product Line
Cascade Biologics™
Quantity
2 Preps
Shipping Condition
Dry Ice
Species
Human

Contents & Storage

minus 20 degrees C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text