Smooth Muscle Growth Supplement (SMGS)
Smooth Muscle Growth Supplement (SMGS)
Gibco™

Smooth Muscle Growth Supplement (SMGS)

Smooth Muscle Growth Supplement (SMGS) is a sterile, concentrated (20X) solution intended for use with Medium 231 to culture humanRead more
Have Questions?
Catalog NumberQuantity
S0072525 mL
Catalog number S00725
Price (USD)
147.65
Online Exclusive
155.00
Save 7.35 (5%)
Each
-
Add to cart
Quantity:
25 mL
Price (USD)
147.65
Online Exclusive
155.00
Save 7.35 (5%)
Each
Add to cart
Ask our AI about this Product
Smooth Muscle Growth Supplement (SMGS) is a sterile, concentrated (20X) solution intended for use with Medium 231 to culture human smooth muscle cells. Each 25-mL bottle of SMGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human smooth muscle cells and is the correct amount of supplement for a 500-mL bottle of basal Medium 231.

When a 500-mL bottle of medium is supplemented with SMGS, the final concentrations of the components in the supplemented medium are:

• Fetal bovine serum: 4.9% v/v
• Human basic fibroblast growth factor: 2 ng/mL
• Human epidermal growth factor: 0.5 ng/mL
• Heparin: 5 ng/mL
• Recombinant human insulin-like growth factor-I: 0.01 μg/mL
• BSA: 0.2 μg/mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeSmooth Muscle Cells
Concentration20 X
Product LineCascade Biologics™
Quantity25 mL
Shipping ConditionDry Ice
SpeciesHuman
Product TypeGrowth Supplement
SterilitySterile
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C).
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link