EpiLife™ Defined Growth Supplement (EDGS)
EpiLife™ Defined Growth Supplement (EDGS)
Gibco™

EpiLife™ Defined Growth Supplement (EDGS)

A defined, sterile, concentrated (100X) solution intended for use with EpiLife™ medium to culture human epidermal keratinocytes. EDGS is notRead more
Have Questions?
Catalog number S0125
Price (USD)/ 5 mL
78.75
-
Add to cart
Price (USD)/ 5 mL
78.75
Add to cart
EpiLife™ Defined Growth Supplement (EDGS)
Catalog numberS0125
Price (USD)/ 5 mL
78.75
-
Add to cart
A defined, sterile, concentrated (100X) solution intended for use with EpiLife™ medium to culture human epidermal keratinocytes. EDGS is not intended for use with Medium 154. EDGS contains all of the growth factors and hormones necessary for the growth of human epidermal keratinocytes. Each bottle contains the correct amount of supplement for a 500 ml bottle of EpiLife™ medium. EDGS contains:
• purified bovine serum albumin
• purified bovine transferrin
• hydrocortisone
• recombinant human insulin-like growth factor type-1 (rhIGF-1)
• prostaglandin E2 (PGE2)
• recombinant human epidermal growth factor (rhEGF) The bovine products in EDGS have been isolated from animals of North American origin. The isolation process for these components includes steps that inactivate viruses. All components of EDGS are greater than 95% pure.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeKeratinocyte Cells
Concentration100 X
Product TypeGrowth Supplement
SterilitySterile
Product LineCascade Biologics™, EpiLife™
Quantity5 mL
Shipping ConditionDry Ice
SpeciesHuman
Unit Size5 mL
Contents & Storage
minus 20 degrees C