QuEChERS Original 2003 Method Extraction Kits
QuEChERS Original 2003 Method Extraction Kits
Thermo Scientific™

QuEChERS Original 2003 Method Extraction Kits

Use Thermo Scientific™ QuEChERS Original 2003 Method Extraction Kits to analyze fruits and vegetables for pesticides, veterinary drug residues, and toxins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionIncludesSalt CompositionSample Size
S1-10-ORIG-POTQuEChERS Original Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl in Pouches; 50/Pk.Salts in Pouches4 g MgSO4, 1 g NaCl10 g sample
S1-10-ORIG-CH-KITQuEChERS Original Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl in Pouches; 50 mL Tubes; Ceramic Homogenizers; 50/Pk.Salts in Pouches, 50 mL Tubes, Ceramic Homogenizers4 g MgSO4, 1 g NaCl10 g sample
S1-10-ORIG-KITQuEChERS Original Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl in Pouches; 50 mL Tubes; 50/Pk.Salts in Pouches, 50 mL Tubes4 g MgSO4, 1 g NaCl10 g sample
S1-15-ORIG-CH-KITQuEChERS Original Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaCl in Pouches; 50 mL Tubes; Ceramic Homogenizers; 50/Pk.Salts in Pouches, 50 mL Tubes, Ceramic Homogenizers6 g MgSO4, 1.5 g NaCl15 g sample
S1-15-ORIG-POTQuEChERS Original Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaCl in Pouches; 50/Pk.Salts in Pouches6 g MgSO4, 1.5 g NaCl15 g sample
S1-15-ORIG-KITQuEChERS Original Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaCl in Pouches; 50 mL Tubes; 50/Pk.Salts in Pouches, 50 mL Tubes6 g MgSO4, 1.5 g NaCl15 g sample
Catalog number S1-10-ORIG-POT
Price (USD)
170.34
Pack of 50
Add to cart
Description:
QuEChERS Original Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl in Pouches; 50/Pk.
Includes:
Salts in Pouches
Salt Composition:
4 g MgSO4, 1 g NaCl
Sample Size:
10 g sample
Price (USD)
170.34
Pack of 50
Add to cart
Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis for pesticide residues, veterinary drug residues, and toxins using the Original 2003 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

Thermo Scientific™ QuEChERS Original 2003 Method Extraction Kits make sample preparation straight-forward and efficient. With our QuEChERS kits, you can confidently perform extractions and analyze fruit and vegetable samples for pesticide residues and other contaminants.

Three different extraction kit types are available to support the Original 2003 QuEChERS Method.

  • Extraction Salts in Pouches, no Tubes – for 10 g or 15 g samples
  • Extraction Salts in Pouches with 50 mL Tubes – for 10 g or 15 g samples
  • Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers – for 10 g or 15 g samples

What is the purpose of the chemicals used in the Original 2003 Method?

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning step; removes remaining water from sample during dispersive solid phase extraction (dSPE) step
  • NaCl: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning step; removes residual water from sample; reduces polar interference and enhances extraction
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample during dispersive solid phase extraction (dSPE) step
Specifications
DescriptionQuEChERS Original Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl in Pouches; 50/Pk.
IncludesSalts in Pouches
MethodQuEChERS sample preparation for Original 2003 Method
Salt Composition4 g MgSO4, 1 g NaCl
Sample Size10 g sample
Sample TypeAll
TypeQuEChERS Extraction Kit
Unit SizePack of 50

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link