Invitrogen™

SYPRO™ Ruby Protein Blot Stain

Catalog number: S11791
Have Questions?
Invitrogen™

SYPRO™ Ruby Protein Blot Stain

Catalog number: S11791
Have Questions?
Catalog Number
S11791
Unit Size
200 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S11791200 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
SYPRO Ruby Protein Blot Stain is a highly sensitive, ready-to-use fluorescent stain for proteins on nitrocellulose or PVDF membranes, making it easy to assess the efficiency of protein transfer to the blot and to determine if lanes are loaded equally. It is compatible with immunostaining (western blotting), microsequencing, and mass spectrometry. Stained proteins can be viewed with blue-LED or UV transilluminator, or with imaging equipment with appropriate filters or laser.

Compare all fluorescent stains ›
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Blot Detection
Detection Method
Fluorescent
Label or Dye
SYPRO Ruby
Product Type
Protein Blot Stain
Product Line
SYPRO™
Quantity
200 mL
Shipping Condition
Room Temperature
Target Molecule
Protein

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text