Sulforhodamine B is a water-soluble, nonfixable red fluorescent (excitation/emission maxima ∼ 565/586 nm) polar tracer that can be used forRead more
Have Questions?
Catalog Number
Quantity
S1307
5 g
Catalog number S1307
Price (KRW)
768,000
Online offer
Ends: 30-Jun-2025
903,000
Save 135,000 (15%)
Each
Add to cart
Quantity:
5 g
Price (KRW)
768,000
Online offer
Ends: 30-Jun-2025
903,000
Save 135,000 (15%)
Each
Add to cart
Sulforhodamine B is a water-soluble, nonfixable red fluorescent (excitation/emission maxima ∼ 565/586 nm) polar tracer that can be used for investigating neuronal morphology, preparing fluorescent liposomes, studying cell-cell communications and following phagosome-lysosome uptake and fusion.
For Research Use Only. Not for use in diagnostic procedures.