Streptavidin, Alexa Fluor™ 633 conjugate
Streptavidin, Alexa Fluor™ 633 conjugate
Invitrogen™

Streptavidin, Alexa Fluor™ 633 conjugate

Alexa Fluor™ 633 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin hasRead more
Have Questions?
Catalog number S21375
Price (USD)/ 1 mg
302.65
Online exclusive
309.00 
Save 6.35 (2%)
-
Add to cart
Price (USD)/ 1 mg
302.65
Online exclusive
309.00 
Save 6.35 (2%)
Add to cart
Streptavidin, Alexa Fluor™ 633 conjugate
Catalog numberS21375
Price (USD)/ 1 mg
302.65
Online exclusive
309.00 
Save 6.35 (2%)
-
Add to cart
Alexa Fluor™ 633 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.

Important Features of Alexa Fluor™ 633 Streptavidin Conjugates:
Alexa Fluor™ 633 streptavidin conjugate has Ex/Em maxima of ∼ (632/647)
Bright, photostable fluorescence
High solubility in aqueous solutions
Available in multiple colors
Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

Properties of Alexa Fluor™ Dyes
Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.

Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Related Links:

Learn more about Avidin-Biotin Detection

Learn more about Alexa Fluor™ Dyes

Find out about other Labeled Streptavidin Conjugates

Read Avidin and Streptavidin Conjugates-Section 7.6 in the Molecular Probes Handbook
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateAlexa Fluor 633
Quantity1 mg
Shipping ConditionRoom Temperature
FormSolid
Product LineAlexa Fluor™
Product TypeStreptavidin Conjugate (fluorescent)
Label or DyeAlexa Fluor Dyes
Unit Size1 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.