Invitrogen™

Streptavidin, R-Phycoerythrin Conjugate (SAPE) - premium grade - 1 mg/mL

Catalog number: S21388
Have Questions?
Invitrogen™

Streptavidin, R-Phycoerythrin Conjugate (SAPE) - premium grade - 1 mg/mL

Catalog number: S21388
Have Questions?
Catalog Number
S21388
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S213881 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The R-phycoerythrin (R-PE) conjugate of streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (R-phycoerythrin). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Streptavidin R-PE is very valuable in biotin-streptavidin detection protocols developed for flow cytometry, microplate assays, and microarrays. This R-PE streptavidin conjugate is a high-purity product (purity confirmed by HPLC) and is supplied as 1 mL of a 1 mg/mL solution.

Important Features of Streptavidin RPE:
Ex/Em maxima of ∼ (496, 546, 565/578) nm
5 to 10 times brighter than organic dyes
Very high solubility in aqueous solutions
Free of all unconjugated streptavidin; and mostly free of unconjugated R-PE
Ideal for flow cytometry, microplate assays, and microarrays

Properties of R-phycoerythrin
R-PE has a quantum efficiency of 78%, making it an incredibly bright reagent, useful where high sensitivity is needed. Because of poor photostability, however, it is not recommended for fluorescence microscopy.

Find Biotinylated Conjugates
We offer an extensive line of biotinylated conjugates for use in biotin-streptavidin detection strategies.
• Use the primary antibody search tool to find biotinylated primary antibodies
• Use the secondary antibody selector tool to find biotinylated secondary antibodies and biotinylated anti-dye and anti-hapten antibodies

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Related Links:

Learn more about Avidin-Biotin Detection

Learn more about the practical applications for Streptavidin R-PE
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
1 mg/mL
Conjugate
R-phycoerythrin
Shipping Condition
Wet Ice
Form
Liquid
Product Type
Streptavidin Conjugate (fluorescent)

Contents & Storage

Store in refrigerator (2–8°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text