AcroMetrix EDTA Plasma Dilution Matrix
Product Image
Thermo Scientific™
AcroMetrix EDTA Plasma Dilution Matrix
Prepare specimens for testing in plasma-based nucleic acid assays with the Thermo Scientific™ AcroMetrix™ EDTA Plasma Dilution Matrix, which is for Research Use Only.
Have Questions?
Catalog NumberQuantity
S228445 mL
Catalog number S2284
Price (USD)
-
Request A Quote
Quantity:
45 mL
Ask our AI about this Product
Prepare specimens for testing in plasma-based nucleic acid assays with the Thermo Scientific™ AcroMetrix™ EDTA Plasma Dilution Matrix (RUO). This matrix is a human plasma matrix shown to be non-reactive for blood borne pathogens, viral nucleic acids and serological markers. It is not intended for use as a substitute for the integral diluents, standards or controls provided by in vitro diagnostic kit manufacturers. The matrix is for Research Use Only and is not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeDilution Matrix
Packaging45mL
Quantity45 mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link