SNARF™-4F 5-(and-6)-Carboxylic Acid
SNARF™-4F 5-(and-6)-Carboxylic Acid
Invitrogen™

SNARF™-4F 5-(and-6)-Carboxylic Acid

The cell-impermeant ratiometric pH indicator SNARF™-4F 5-(and-6)-carboxylic acid (pKa of ∼6.4) exhibits a significant pH-dependent emission shift from yellow-orange toRead more
Have Questions?
Catalog NumberQuantity
S239201 mg
Catalog number S23920
Price (USD)
508.00
Each
-
Add to cart
Quantity:
1 mg
Price (USD)
508.00
Each
Add to cart
Ask our AI about this Product
The cell-impermeant ratiometric pH indicator SNARF™-4F 5-(and-6)-carboxylic acid (pKa of ∼6.4) exhibits a significant pH-dependent emission shift from yellow-orange to deep red fluorescence under acidic and basic conditions, respectively. This pH dependence allows the ratio of the fluorescence intensities from the dye at two emission wavelengths - typically 580 nm and 640 nm - to be used for quantitative determinations of pH.

Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeFluorescent Dye-Based
IndicatorpH Indicator
Quantity1 mg
Shipping ConditionRoom Temperature
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Microplate Reader
Product LineSNARF
Product TypepH Indicator
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link